4-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((2-Fluorophenyl)carbamoyl)phenyl)boronic acid

A739738-1G A739738-250MG A739738-5G
76879-936EA 55.17 USD
76879-936 76879-938 76879-940
4-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(2-Fluorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
MW: 259.0408 g/mol
MDL Number: MFCD09027205
CAS Number: 874288-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR