7-Isoquinolineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-7-ylboronic acid

A739356-1G A739356-250MG A739356-5G A739356-100MG
76879-930EA 160.2 USD
76879-930 76879-932 76879-934 77674-034
7-Isoquinolineboronic acid 98%
7-Isoquinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
MW: 172.9763 g/mol
MDL Number: MFCD08234617
CAS Number: 1092790-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR