2-Hydroxypyridine-4-boronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Boronopyridin-2-ol, (2-Hydroxypyridin-4-yl)boronic acid

A738644-100MG A738644-250MG A738644-1G
76879-926EA 40 USD
76879-926 76879-928 77884-658
2-Hydroxypyridine-4-boronic acid 98+%
2-Hydroxypyridine-4-boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BNO₃
MDL Number: MFCD11223187
CAS Number: 902148-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR