2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-5-methylthiazole-4-carboxylate

A737625-100MG A737625-1G A737625-250MG A737625-5G
76879-918EA 20.16 USD
76879-918 76879-920 76879-922 76879-924
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 97%
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂S
MW: 205.66 g/mol
MDL Number: MFCD19443895
CAS Number: 907545-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR