Diethyl ((methylsulfonyl)methyl)phosphonate 95%

Supplier: Ambeed
Warning

A736416-1G A736416-5G
76879-916EA 129.43 USD
76879-916 77552-266
Diethyl ((methylsulfonyl)methyl)phosphonate 95%
Diethyl ((methylsulfonyl)methyl)phosphonate

Precaución: for Research Use Only

Formula: C₆H₁₅O₅PS
MDL Number: MFCD00181919
CAS Number: 40137-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR