2-Bromo-2'-chloro-6'-fluoroacetophenone 98%

Supplier: Ambeed
Danger

A735802-100MG A735802-1G A735802-250MG
76879-910EA 15 USD
76879-910 76879-912 76879-914
2-Bromo-2'-chloro-6'-fluoroacetophenone 98%
2-Bromo-2'-chloro-6'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrClFO
MDL Number: MFCD07368692
CAS Number: 157359-99-4
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR