2-(4-Bromo-3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A734158-1G A734158-250MG A734158-5G
76879-902EA 91.24 USD
76879-902 76879-904 76879-906
2-(4-Bromo-3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-Bromo-3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BBrCl₂O₂
MW: 351.85 g/mol
MDL Number: MFCD12405328
CAS Number: 942069-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR