5-Norbornene-2-carbonitrile (mixture of isomers) 98% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carbonitrile, 5-Norbornene-2-carbonitrile, 5-Cyanobicyclo[2.2.1]hept-2-ene, 5-Cyano-2-norbornene

A732629-100G A732629-25G A732629-5G
76879-888EA 174 USD
76879-888 76879-890 76879-892
5-Norbornene-2-carbonitrile (mixture of isomers) 98% (mixture of isomers)
5-Norbornene-2-carbonitrile (mixture of isomers)

Precaución: for Research Use Only

Formula: C₈H₉N
MDL Number: MFCD00167563
CAS Number: 95-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR