(R)-8-(Benzyloxy)-5-(oxiran-2-yl)quinolin-2(1H)-one 98+%

Supplier: Ambeed
Warning

A731237-10G A731237-5G
76879-884EA 280.08 USD
76879-884 76879-886
(R)-8-(Benzyloxy)-5-(oxiran-2-yl)quinolin-2(1H)-one 98+%
(R)-8-(Benzyloxy)-5-(oxiran-2-yl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₃
MW: 293.31 g/mol
MDL Number: MFCD17018704
CAS Number: 173140-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR