5-Amino-4-cyano-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A730318-100MG A730318-250MG A730318-1G
76879-880EA 42 USD
76879-880 76879-882 77884-647
5-Amino-4-cyano-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole 98%
5-Amino-4-cyano-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₈F₂N₄
MDL Number: MFCD08459254
CAS Number: 1020057-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR