6-Bromo-2-naphthaldehyde 98%

Supplier: Ambeed
Warning

A730287-100MG A730287-1G A730287-250MG A730287-5G
76879-872EA 51.99 USD
76879-872 76879-874 76879-876 76879-878
6-Bromo-2-naphthaldehyde 98%
6-Bromo-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇BrO
MW: 235.08 g/mol
MDL Number: MFCD09842469
CAS Number: 170737-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR