8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride 97+%

Supplier: Ambeed
Warning

A729134-10G A729134-1G
76879-864EA 58 USD
76879-864 76879-866
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride 97+%
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO·ClH
MW: 251.752 g/mol
MDL Number: MFCD06659471
CAS Number: 83393-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR