(5-Fluoro-2-(N-methylsulfamoyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A726134-1G A726134-250MG A726134-5G
76879-848EA 202.64 USD
76879-848 76879-850 76879-852
(5-Fluoro-2-(N-methylsulfamoyl)phenyl)boronic acid 98%
(5-Fluoro-2-(N-methylsulfamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BFNO₄S
MW: 233.03 g/mol
MDL Number: MFCD15143479
CAS Number: 1218790-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR