3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%

Supplier: Ambeed
Danger

Synonyms: (3-Chloro-4-(cyclohexylcarbamoyl)phenyl)boronic acid

A723820-1G A723820-250MG A723820-5G
76879-842EA 64 USD
76879-842 76879-844 76879-846
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClNO₃
MDL Number: MFCD09475818
CAS Number: 957034-65-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR