3,5-Dichloro-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A723641-1G A723641-5G
76879-838EA 61.54 USD
76879-838 76879-840
3,5-Dichloro-4-fluorobenzaldehyde 98%
3,5-Dichloro-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MW: 193.0000 g/mol
MDL Number: MFCD11845864
CAS Number: 117820-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR