(2,3-Difluoro-4-isopropoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Difluoro-4-isopropoxyphenylboronic acid

A723415-1G A723415-250MG A723415-5G
76879-832EA 161.26 USD
76879-832 76879-834 76879-836
(2,3-Difluoro-4-isopropoxyphenyl)boronic acid 98%
(2,3-Difluoro-4-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
MDL Number: MFCD19237183
CAS Number: 1154762-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR