5-Bromo-3-fluoro-2-nitrotoluene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-1-fluoro-3-methyl-2-nitrobenzene
A722634-100MG
A722634-250MG
A722634-5G
A722634-1G
76879-828EA
11
USD
76879-828
76879-830
77569-372
77614-226
5-Bromo-3-fluoro-2-nitrotoluene 98%
5-Bromo-3-fluoro-2-nitrotoluene
Precaución: For research use only.
Formula:
C₇H₅BrFNO₂ |
MDL Number:
MFCD22566379 CAS Number: 1224629-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...