3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzoate

A722212-250MG A722212-5G
76879-824EA 26 USD
76879-824 76879-826
3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 98%
3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₅
MW: 278.11 g/mol
MDL Number: MFCD08458200
CAS Number: 1073371-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR