1-Bromo-3-(difluoromethoxy)-5-fluorobenzene 97%

Supplier: Ambeed
Danger

A720006-100MG A720006-1G A720006-250MG
76879-810EA 31.83 USD
76879-810 76879-812 76879-814
1-Bromo-3-(difluoromethoxy)-5-fluorobenzene 97%
1-Bromo-3-(difluoromethoxy)-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
MDL Number: MFCD18390899
CAS Number: 433939-28-7
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR