4'-Acetylbiphenyl-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4'-Acetyl-[1,1'-biphenyl]-4-yl)boronic acid

A719910-100MG A719910-1G A719910-250MG
76879-804EA 40 USD
76879-804 76879-806 76879-808
4'-Acetylbiphenyl-4-boronic acid 98%
4'-Acetylbiphenyl-4-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BO₃
MDL Number: MFCD16295057
CAS Number: 1029438-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR