6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate

A719363-100MG A719363-1G A719363-250MG
76879-798EA 29 USD
76879-798 76879-800 76879-802
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 98%
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₄
MDL Number: MFCD09037484
CAS Number: 1032758-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR