PHA-665752 99%

Supplier: Ambeed
Warning

Synonyms: 2R)-1-[[5-[(Z)-[5-[[(2,6-Dichlorophenyl)methyl]sulphonyl]-1,2-dihydro-2-oxo-3H-indol-3-ylidene]methyl]-2,4-dimethyl-1H-pyrrol-3-yl]carbonyl]-2-(1-pyrrolidinylmethyl)pyrrolidine, (R,Z)-5-((2,6-Dichlorobenzyl)sulphonyl)-3-((3,5-dimethyl-4-(2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl)-1H-pyrrol-2-yl)methylene)indolin-2-one

A717441-10MG A717441-1MG A717441-5MG
76879-792EA 146 USD
76879-792 76879-794 76879-796
PHA-665752 99%
PHA-665752

Precaución: for Research Use Only

Formula: C₃₂H₃₄Cl₂N₄O₄S
MW: 641.61 g/mol
MDL Number: MFCD07772270
CAS Number: 477575-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR