Propargyl benzoate 95%

Supplier: Ambeed
Warning

Synonyms: Prop-2-yn-1-yl benzoate

A717349-100G A717349-1G A717349-250MG A717349-25G A717349-5G
76879-782EA 927.23 USD
76879-782 76879-784 76879-786 76879-788 76879-790
Propargyl benzoate 95%
Propargyl benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00996341
CAS Number: 6750-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR