4-((Morpholino)methyl)phenylboronic acid pinacol ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine hydrochloride

A717159-1G A717159-5G A717159-250MG
76879-776EA 33 USD
76879-776 76879-778 77886-652
4-((Morpholino)methyl)phenylboronic acid pinacol ester hydrochloride 97%
4-((Morpholino)methyl)phenylboronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₃·ClH
MDL Number: MFCD06657894
CAS Number: 944591-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR