1-Ethynyl-2,4-difluorobenzene 95%

Supplier: Ambeed
Danger

A716110-100MG A716110-1G A716110-250MG A716110-5G
76879-768EA 26 USD
76879-768 76879-770 76879-772 76879-774
1-Ethynyl-2,4-difluorobenzene 95%
1-Ethynyl-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₄F₂
MDL Number: MFCD02093726
CAS Number: 302912-34-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR