3-(Aminomethyl)-4,6-dimethylpyridin-2(1H)-one hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A714837-100G
A714837-10G
A714837-1G
A714837-250MG
A714837-25G
A714837-5G
76879-744EA
332
USD
76879-744
76879-746
76879-748
76879-750
76879-752
76879-754
3-(Aminomethyl)-4,6-dimethylpyridin-2(1H)-one hydrochloride 95%
3-(Aminomethyl)-4,6-dimethylpyridin-2(1H)-one hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O·ClH |
MDL Number:
MFCD09053337 CAS Number: 1173081-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...