3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-4-cyclopentylcarbamoylphenylboronic acid, (3-Chloro-4-(cyclopentylcarbamoyl)phenyl)boronic acid

A713945-1G A713945-250MG A713945-5G
76879-738EA 60.48 USD
76879-738 76879-740 76879-742
3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₃
MW: 267.5200 g/mol
MDL Number: MFCD09258664
CAS Number: 957061-19-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR