4-(Chloromethyl)benzonitrile 98%
Supplier: Ambeed
|
Danger
|
A713677-100G
A713677-25G
A713677-500G
76879-730EA
19
USD
76879-730
76879-732
76879-734
76879-736
4-(Chloromethyl)benzonitrile 98%
4-(Chloromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆ClN |
MDL Number:
MFCD00019761 CAS Number: 874-86-2 UN: 2923 ADR: 8,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...