2-((4-Methoxybenzyl)amino)ethanol 95%

Supplier: Ambeed
Warning

A713508-100MG A713508-1G A713508-250MG A713508-5G A713508-10G
76879-722EA 35 USD
76879-722 76879-724 76879-726 76879-728 77884-628
2-((4-Methoxybenzyl)amino)ethanol 95%
2-((4-Methoxybenzyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD05863671
CAS Number: 64834-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR