2-(5-Bromo-2-fluorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A710853-10G A710853-1G A710853-5G
76879-690EA 47 USD
76879-690 76879-692 76879-694
2-(5-Bromo-2-fluorophenyl)acetonitrile 98%
2-(5-Bromo-2-fluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.037 g/mol
MDL Number: MFCD09804785
CAS Number: 305800-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR