N-(5-Bromo-4-ethylthiazol-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A710476-100MG A710476-250MG
76879-686EA 113.52 USD
76879-686 76879-688
N-(5-Bromo-4-ethylthiazol-2-yl)acetamide 98%
N-(5-Bromo-4-ethylthiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₉BrN₂OS
MW: 249.13 g/mol
MDL Number: MFCD12756845
CAS Number: 1267904-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR