Boc-4-(trifluoromethyl)-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)-D-phenylalanine, N-BOC protected, (R)-2-((tert-Butoxycarbonyl)amino)-3-(4-(trifluoromethyl)phenyl)propanoic acid

A707258-1G A707258-250MG A707258-5G
76879-662EA 59 USD
76879-662 76879-664 76879-666
Boc-4-(trifluoromethyl)-D-phenylalanine 97%
Boc-4-(trifluoromethyl)-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MW: 333.31 g/mol
MDL Number: MFCD00797557
CAS Number: 82317-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR