2-(Oxetan-3-yl)ethanol 98%

Supplier: Ambeed
Danger

A706195-250MG A706195-1G A706195-100MG A706195-5G
76879-654EA 24 USD
76879-654 77614-176 77886-643 77886-644
2-(Oxetan-3-yl)ethanol 98%
2-(Oxetan-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD17215859
CAS Number: 251922-46-0
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR