4-(4-Aminophenoxy)benzonitrile 98%

Supplier: Ambeed
Warning

A703873-250MG A703873-5G
76879-638EA 39 USD
76879-638 76879-640
4-(4-Aminophenoxy)benzonitrile 98%
4-(4-Aminophenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O
MDL Number: MFCD00197201
CAS Number: 17076-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR