Dibenzo[b,d]furan-2-ol 95%

Supplier: Ambeed
Warning

A703749-100G A703749-1G A703749-25G A703749-5G
76879-630EA 296 USD
76879-630 76879-632 76879-634 76879-636
Dibenzo[b,d]furan-2-ol 95%
Dibenzo[b,d]furan-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MDL Number: MFCD00010325
CAS Number: 86-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR