(4-Chloro-3-ethylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A703647-100MG A703647-1G A703647-250MG A703647-5G
76879-622EA 12 USD
76879-622 76879-624 76879-626 76879-628
(4-Chloro-3-ethylphenyl)boronic acid 98%
(4-Chloro-3-ethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₂
MDL Number: MFCD10566599
CAS Number: 918810-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR