(2-(Benzyloxy)-5-(trifluoromethyl)pyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

A703362-100MG A703362-1G A703362-250MG A703362-5G
76879-614EA 36.08 USD
76879-614 76879-616 76879-618 76879-620
(2-(Benzyloxy)-5-(trifluoromethyl)pyridin-3-yl)boronic acid 98%
(2-(Benzyloxy)-5-(trifluoromethyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₃NO₃
MW: 297.04 g/mol
MDL Number: MFCD13195242
CAS Number: 850864-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR