Sorbitan trioleate Saponification Value 165-180

Supplier: Ambeed
Warning

A702269-1000G A702269-500G A702269-100G
76879-610EA 19 USD
76879-610 76879-612 77884-606
Sorbitan trioleate Saponification Value 165-180
Sorbitan trioleate

Precaución: for Research Use Only

Formula: C₆₀H₁₀₈O₈
MDL Number: MFCD00133820
CAS Number: 26266-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR