4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Propyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Propylureido)benzeneboronic acid, pinacol ester

A696895-1G A696895-250MG A696895-5G
76879-602EA 108 USD
76879-602 76879-604 76879-606
4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BN₂O₃
MDL Number: MFCD09027288
CAS Number: 874291-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR