Ethyl 3-hydroxypicolinate 98%

Supplier: Ambeed
Warning

A696722-1G A696722-250MG
76879-598EA 26.53 USD
76879-598 76879-600
Ethyl 3-hydroxypicolinate 98%
Ethyl 3-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
MDL Number: MFCD11656384
CAS Number: 73406-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR