(S)-3-(Boc-amino)-3-(3-(trifluoromethyl)phenyl)propionic acid 95%

Supplier: Ambeed
Warning

A693444-100MG A693444-1G A693444-250MG A693444-5G
76879-588EA 9 USD
76879-588 76879-590 76879-592 76879-594
(S)-3-(Boc-amino)-3-(3-(trifluoromethyl)phenyl)propionic acid 95%
(S)-3-(Boc-amino)-3-(3-(trifluoromethyl)phenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MDL Number: MFCD03427926
CAS Number: 500770-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR