4-(Difluoromethoxy)-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A690695-1G A690695-5G
76879-576EA 42 USD
76879-576 77552-100
4-(Difluoromethoxy)-2-fluorobenzaldehyde 98%
4-(Difluoromethoxy)-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD11847162
CAS Number: 1214358-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR