(3-(Morpholine-4-carbonyl)-5-nitrophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A690540-1G A690540-250MG A690540-5G
76879-570EA 124.13 USD
76879-570 76879-572 76879-574
(3-(Morpholine-4-carbonyl)-5-nitrophenyl)boronic acid 98%
(3-(Morpholine-4-carbonyl)-5-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BN₂O₆
MW: 280.04 g/mol
MDL Number: MFCD08056359
CAS Number: 871332-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR