Ethyl 5-bromo-1-methyl-1H-imidazole-2-carboxylate 95%

Supplier: Ambeed
Warning

A687563-100MG A687563-1G A687563-250MG
76879-560EA 92.3 USD
76879-560 76879-562 76879-564
Ethyl 5-bromo-1-methyl-1H-imidazole-2-carboxylate 95%
Ethyl 5-bromo-1-methyl-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O₂
MW: 233.06 g/mol
MDL Number: MFCD18249878
CAS Number: 865798-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR