6-Bromo-2,2-dimethylchroman-4-one 98%

Supplier: Ambeed
Warning

A687419-10G A687419-1G A687419-250MG A687419-5G
76879-552EA 92.3 USD
76879-552 76879-554 76879-556 76879-558
6-Bromo-2,2-dimethylchroman-4-one 98%
6-Bromo-2,2-dimethylchroman-4-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.10 g/mol
MDL Number: MFCD12068434
CAS Number: 99853-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR