3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 3-Chloro-5-(piperidin-1-carbonyl)phenylboronic acid

A686800-1G A686800-250MG A686800-5G
76879-546EA 95 USD
76879-546 76879-548 76879-550
3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid 98%
3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₃
MDL Number: MFCD09800859
CAS Number: 957120-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR