3-((5-Bromopyridin-2-yl)oxy)-N,N-dimethylpropan-1-amine 95%

Supplier: Ambeed
Warning

A686250-1G A686250-25G A686250-5G
76879-540EA 30 USD
76879-540 76879-542 76879-544
3-((5-Bromopyridin-2-yl)oxy)-N,N-dimethylpropan-1-amine 95%
3-((5-Bromopyridin-2-yl)oxy)-N,N-dimethylpropan-1-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD14652180
CAS Number: 1248399-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR