3,3'-(Methylazanediyl)bis(propan-1-ol) 95%

Supplier: Ambeed
Warning

A684367-100MG A684367-250MG A684367-1G
76879-536EA 103.97 USD
76879-536 76879-538 77614-118
3,3'-(Methylazanediyl)bis(propan-1-ol) 95%
3,3'-(Methylazanediyl)bis(propan-1-ol)

Precaución: for Research Use Only

Formula: C₇H₁₇NO₂
MW: 147.21 g/mol
MDL Number: MFCD29919632
CAS Number: 2158-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR