1-Bromo-4-ethyl-2-fluorobenzene 95%
Supplier: Ambeed
|
Warning
|
A683576-100MG
A683576-250MG
A683576-1G
76879-532EA
77
USD
76879-532
76879-534
77884-579
1-Bromo-4-ethyl-2-fluorobenzene 95%
1-Bromo-4-ethyl-2-fluorobenzene
Precaución: for Research Use Only
Formula:
C₈H₈BrF |
MDL Number:
MFCD22044817 CAS Number: 928304-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...