(R)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A681574-100MG A681574-1G A681574-250MG
76879-526EA 25 USD
76879-526 76879-528 76879-530
(R)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 98%
(R)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD06203312
CAS Number: 660852-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR